Assay Details
| Gene Symbol: DDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115878.2 | XP_014971364.1 | 873 | 77 | ||
| XM_015115879.2 | XP_014971365.1 | 872 | 77 | |||
| XM_015115880.2 | XP_014971366.1 | 1552 | 77 | |||
| GenBank mRNA | JU333199.1 | - | 910 | 77 | ||
| JU474512.1 | - | 916 | 77 | |||
| JV046323.1 | - | 864 | 77 | |||
More Information
Set Membership: |
|

DDX6
Genomic Map