Assay Details
| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099347.4 | XP_001099347.2 | 844 | 78 | ||
| GenBank mRNA | JU328454.1 | - | 170 | 78 | ||
| JV723209.1 | - | 274 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHAC1| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099347.4 | XP_001099347.2 | 844 | 78 | ||
| GenBank mRNA | JU328454.1 | - | 170 | 78 | ||
| JV723209.1 | - | 274 | 78 | |||