Assay Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257822.1 | NP_001244751.1 | 426 | 71 | ||
| XM_028852177.1 | XP_028708010.1 | 676 | 71 | |||
| GenBank mRNA | JU337331.1 | - | 426 | 71 | ||
| JU478031.1 | - | 422 | 71 | |||
| JV048834.1 | - | 374 | 71 | |||
| JV601635.1 | - | 427 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTDSS1
Genomic Map