Assay Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099023.4 | XP_001099023.2 | 397 | 81 | ||
| XM_028846058.1 | XP_028701891.1 | 397 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TFPI2
Genomic Map