Assay Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144073.2 | XP_014999559.2 | 2246 | 97 | ||
| XM_015144075.2 | XP_014999561.1 | 2030 | 97 | |||
| XM_015144076.2 | XP_014999562.1 | 2222 | 97 | |||
| XM_015144077.2 | XP_014999563.1 | 2003 | 97 | |||
| XM_028851661.1 | XP_028707494.1 | 2122 | 97 | |||
| GenBank mRNA | JU322082.1 | - | 1512 | 97 | ||
| JU470890.1 | - | 1587 | 97 | |||
| JV044479.1 | - | 1587 | 97 | |||
| JV489169.1 | - | 1501 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF410
Genomic Map