Assay Details
| Gene Symbol: SLC39A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151984.2 | XP_015007470.1 | 1682 | 90 | ||
| XM_015151985.2 | XP_015007471.1 | 1695 | 90 | |||
| XM_015151988.2 | XP_015007474.1 | 1668 | 90 | |||
| XM_015151989.2 | XP_015007475.1 | 1663 | 90 | |||
| XM_015151990.2 | XP_015007476.1 | 1666 | 90 | |||
| XM_028829609.1 | XP_028685442.1 | 1692 | 90 | |||
| XM_028829610.1 | XP_028685443.1 | 1656 | 90 | |||
More Information
Set Membership: |
|

SLC39A5
Genomic Map