Assay Details
| Gene Symbol: TGFB1I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266172.1 | NP_001253101.1 | 1136 | 58 | ||
| XM_028840577.1 | XP_028696410.1 | 1300 | 58 | |||
| XM_028840578.1 | XP_028696411.1 | 1207 | 58 | |||
| GenBank mRNA | JV636344.1 | - | 1136 | 58 | ||
More Information
Set Membership: |
|

TGFB1I1
Genomic Map