Assay Details
| Gene Symbol: ACD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092222.4 | XP_001092222.3 | 1075 | 76 | ||
| XM_015126428.2 | XP_014981914.1 | 1081 | 76 | |||
| GenBank mRNA | JU491772.1 | - | 553 | 76 | ||
| JV693756.1 | - | 385 | 76 | |||
More Information
Set Membership: |
|

ACD
Genomic Map