Assay Details
| Gene Symbol: JAKMIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257961.1 | NP_001244890.1 | 2141 | 69 | ||
| XM_028848151.1 | XP_028703984.1 | 1981 | 69 | |||
| XM_028848152.1 | XP_028703985.1 | 2192 | 69 | |||
| XM_028848153.1 | XP_028703986.1 | 1981 | 69 | |||
| XM_028848154.1 | XP_028703987.1 | 1981 | 69 | |||
| XM_028848155.1 | XP_028703988.1 | 1981 | 69 | |||
| XM_028848156.1 | XP_028703989.1 | 2192 | 69 | |||
| XM_028848157.1 | XP_028703990.1 | 1981 | 69 | |||
| XM_028848158.1 | XP_028703991.1 | 1981 | 69 | |||
| XM_028848159.1 | XP_028703992.1 | 1981 | 69 | |||
| GenBank mRNA | JU320388.1 | - | 2141 | 69 | ||
| JU471442.1 | - | 1843 | 69 | |||
| JV046278.1 | - | 1855 | 69 | |||
| JV458218.1 | - | 1898 | 69 | |||
More Information
Set Membership: |
|

JAKMIP1
Genomic Map