Assay Details
| Gene Symbol: OTUD7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266823.1 | NP_001253752.1 | 1464 | 68 | ||
| XM_015150531.2 | XP_015006017.1 | 1465 | 68 | |||
| XM_015150550.2 | XP_015006036.1 | 1404 | 68 | |||
| GenBank mRNA | JU322462.1 | - | 1482 | 68 | ||
| JV045692.1 | - | 1464 | 68 | |||
More Information
Set Membership: |
|

OTUD7B
Genomic Map