Assay Details
| Gene Symbol: TM6SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142885.2 | XP_014998371.1 | 792 | 91 | ||
| XM_015142888.2 | XP_014998374.1 | 792 | 91 | |||
| XM_028851246.1 | XP_028707079.1 | 713 | 91 | |||
| XM_028851248.1 | XP_028707081.1 | 792 | 91 | |||
| XM_028851249.1 | XP_028707082.1 | 640 | 91 | |||
| XM_028851250.1 | XP_028707083.1 | 619 | 91 | |||
| XM_028851251.1 | XP_028707084.1 | 639 | 91 | |||
| XM_028851252.1 | XP_028707085.1 | 861 | 91 | |||
| XM_028851253.1 | XP_028707086.1 | 862 | 91 | |||
| XM_028851255.1 | XP_028707088.1 | 845 | 91 | |||
| GenBank mRNA | JU320749.1 | - | 634 | 91 | ||
| JV565964.1 | - | 704 | 91 | |||
| JV619485.1 | - | 210 | 91 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TM6SF1
Genomic Map