Assay Details
| Gene Symbol: WASF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266598.1 | NP_001253527.1 | 677 | 76 | ||
| GenBank mRNA | JU320139.1 | - | 729 | 76 | ||
| JU320140.1 | - | 729 | 76 | |||
| JV044786.1 | - | 677 | 76 | |||
More Information
Set Membership: |
|

WASF3
Genomic Map