Assay Details
| Gene Symbol: SLC1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266066.1 | NP_001252995.1 | 752 | 65 | ||
| XM_015139850.2 | XP_014995336.2 | 1001 | 65 | |||
| GenBank mRNA | JV489183.1 | - | 806 | 65 | ||
| JV564949.1 | - | 786 | 65 | |||
| JV667456.1 | - | 752 | 65 | |||
More Information
Set Membership: |
|

SLC1A3
Genomic Map