Assay Details
| Gene Symbol: NFATC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148778.2 | XP_015004264.1 | 2252 | 53 | ||
| XM_015148780.2 | XP_015004266.1 | 2252 | 53 | |||
| XM_028828511.1 | XP_028684344.1 | 1807 | 53 | |||
| XM_028828512.1 | XP_028684345.1 | 1807 | 53 | |||
More Information
Set Membership: |
|

NFATC2
Genomic Map