Assay Details
| Gene Symbol: TAFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152189.2 | XP_015007675.1 | 1846 | 58 | ||
| XM_015152191.2 | XP_015007677.1 | 1541 | 58 | |||
| XM_015152192.2 | XP_015007678.1 | 281 | 58 | |||
| XM_028829685.1 | XP_028685518.1 | 679 | 58 | |||
| GenBank mRNA | JU320216.1 | - | 638 | 58 | ||
More Information
Set Membership: |
|

TAFA2
Genomic Map