Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032911.1 | NP_001028083.1 | 1960 | 57 | ||
| GenBank mRNA | AY526325.1 | - | 1960 | 57 | ||
| JV650576.1 | - | 203 | 57 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AR
Genomic Map