Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122922.2 | XP_014978408.1 | 1134 | 80 | ||
| GenBank mRNA | JU322107.1 | - | 855 | 80 | ||
| JV045379.1 | - | 859 | 80 | |||
More Information
Set Membership: |
|

ANGPTL1
Genomic Map