Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118380.2 | XP_014973866.2 | 1018 | 72 | ||
| XM_015118381.2 | XP_014973867.2 | 1013 | 72 | |||
| XM_028835939.1 | XP_028691772.1 | 1013 | 72 | |||
| GenBank mRNA | JU323292.1 | - | 72 | |||
| JU473404.1 | - | 72 | ||||
More Information
Set Membership: |
|

ZNF232
Genomic Map