Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119892.2 | XP_014975378.1 | 2250 | 64 | ||
| XM_028836604.1 | XP_028692437.1 | 2370 | 64 | |||
| GenBank mRNA | JV600374.1 | - | 1306 | 64 | ||
| JV613092.1 | - | 714 | 64 | |||
| JV667371.1 | - | 1319 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GFAP
Genomic Map