Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113104.2 | XP_014968590.1 | 654 | 132 | ||
| XM_015113105.2 | XP_014968591.1 | 423 | 132 | |||
| XM_015113107.2 | XP_014968593.1 | 768 | 132 | |||
| XM_015113109.2 | XP_014968595.1 | 432 | 132 | |||
| XM_015113110.2 | XP_014968596.1 | 204 | 132 | |||
| XM_015113111.2 | XP_014968597.1 | 579 | 132 | |||
| XM_015113113.2 | XP_014968599.1 | 608 | 132 | |||
| XM_015113114.2 | XP_014968600.1 | 539 | 132 | |||
| XM_028832290.1 | XP_028688123.1 | 289 | 132 | |||
| XM_028832292.1 | XP_028688125.1 | 422 | 132 | |||
| XM_028832293.1 | XP_028688126.1 | 654 | 132 | |||
| XM_028832294.1 | XP_028688127.1 | 579 | 132 | |||
| GenBank mRNA | JU321498.1 | - | 358 | 132 | ||
| JU470655.1 | - | 167 | 132 | |||
| JV045546.1 | - | 252 | 132 | |||
| JV638942.1 | - | 261 | 132 | |||
More Information
Set Membership: |
|

SEPTIN10
Genomic Map