Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094439.4 | XP_001094439.2 | 783 | 73 | ||
| XM_015126932.2 | XP_014982418.2 | 725 | 73 | |||
| XM_015126933.2 | XP_014982419.2 | 581 | 73 | |||
| GenBank mRNA | JU334239.1 | - | 709 | 73 | ||
| JU475357.1 | - | 648 | 73 | |||
| JV703846.1 | - | 785 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HCCS
Genomic Map