Assay Details
| Gene Symbol: CAPZA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168609.1 | NP_001162080.1 | 256 | 117 | ||
| GenBank mRNA | JU336588.1 | - | 249 | 117 | ||
| JU477402.1 | - | 238 | 117 | |||
| JV048378.1 | - | 188 | 117 | |||
| JV630231.1 | - | 258 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CAPZA2
Genomic Map