Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257519.1 | NP_001244448.1 | 318 | 62 | ||
| GenBank mRNA | JU322576.1 | - | 318 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAXDC2| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257519.1 | NP_001244448.1 | 318 | 62 | ||
| GenBank mRNA | JU322576.1 | - | 318 | 62 | ||
Set Membership: |
|