Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085226.3 | XP_001085226.2 | 428 | 65 | ||
| XM_001085341.3 | XP_001085341.2 | 430 | 65 | |||
| XM_015120223.2 | XP_014975709.1 | 861 | 65 | |||
| GenBank mRNA | JU333313.1 | - | 177 | 65 | ||
| JV046401.1 | - | 214 | 65 | |||
| JV635545.1 | - | 150 | 65 | |||
More Information
Set Membership: |
|

SLC39A11
Genomic Map