Assay Details
| Gene Symbol: SLC35A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106480.4 | XP_001106480.2 | 429 | 92 | ||
| XM_001106663.4 | XP_001106663.1 | 371 | 92 | |||
| XM_028831197.1 | XP_028687030.1 | 429 | 92 | |||
| XM_028831198.1 | XP_028687031.1 | 429 | 92 | |||
| GenBank mRNA | JU333690.1 | - | 374 | 92 | ||
| JU474937.1 | - | 260 | 92 | |||
More Information
Set Membership: |
|

SLC35A3
Genomic Map