Assay Details
| Gene Symbol: PPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260819.1 | NP_001247748.1 | 785 | 76 | ||
| GenBank mRNA | JU335106.1 | - | 785 | 76 | ||
| JV654412.1 | - | 918 | 76 | |||
More Information
Set Membership: |
|

PPAT
Genomic Map