Assay Details
| Gene Symbol: LOC719351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828559.1 | XP_028684392.1 | 654 | 67 | ||
| GenBank mRNA | JU333764.1 | - | 1636 | 67 | ||
| JU475010.1 | - | 1682 | 67 | |||
| JV046633.1 | - | 1683 | 67 | |||
More Information
Set Membership: |
|

LOC719351
Genomic Map