Assay Details
| Gene Symbol: FAXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266112.1 | NP_001253041.1 | 921 | 57 | ||
| GenBank mRNA | JU336750.1 | - | 921 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAXC| Gene Symbol: FAXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266112.1 | NP_001253041.1 | 921 | 57 | ||
| GenBank mRNA | JU336750.1 | - | 921 | 57 | ||
Set Membership: |
|