Assay Details
| Gene Symbol: CNIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193769.1 | NP_001180698.1 | 204 | 132 | ||
| XM_015143701.2 | XP_014999187.1 | 220 | 180 | |||
| GenBank mRNA | JU320593.1 | - | 184 | 132 | ||
| JU472064.1 | - | 155 | 132 | |||
| JV045859.1 | - | 189 | 132 | |||
| JV528696.1 | - | 168 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CNIH1
Genomic Map