Assay Details
| Gene Symbol: SP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266924.1 | NP_001253853.1 | 2091 | 72 | ||
| XM_015151801.2 | XP_015007287.1 | 2019 | 72 | |||
| GenBank mRNA | JU336057.1 | - | 2139 | 72 | ||
| JU476915.1 | - | 2129 | 72 | |||
| JV048023.1 | - | 2091 | 72 | |||
More Information
Set Membership: |
|

SP1
Genomic Map