Assay Details
| Gene Symbol: UGCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257723.1 | NP_001244652.1 | 809 | 69 | ||
| GenBank mRNA | JU320207.1 | - | 809 | 69 | ||
| JU471684.1 | - | 781 | 69 | |||
| JV043883.1 | - | 865 | 69 | |||
More Information
Set Membership: |
|

UGCG
Genomic Map