Assay Details
| Gene Symbol: AURKC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098364.4 | XP_001098364.4 | 1216 | 79 | ||
| GenBank mRNA | JV044723.1 | - | 409 | 79 | ||
| JV636339.1 | - | 470 | 79 | |||
More Information
Set Membership: |
|

AURKC
Genomic Map