Assay Details
| Gene Symbol: ADSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261110.1 | NP_001248039.1 | 366 | 131 | ||
| GenBank mRNA | JU333149.1 | - | 380 | 131 | ||
| JU471846.1 | - | 370 | 131 | |||
| JV046292.1 | - | 366 | 131 | |||
| JV601510.1 | - | 551 | 131 | |||
| JV638577.1 | - | 387 | 131 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADSS2
Genomic Map