Assay Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257602.1 | NP_001244531.1 | 414 | 134 | ||
| GenBank mRNA | JU334516.1 | - | 414 | 134 | ||
| JU334517.1 | - | 414 | 134 | |||
| JV044307.1 | - | 408 | 134 | |||
| JV047063.1 | - | 408 | 134 | |||
| JV581953.1 | - | 421 | 134 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAGOH
Genomic Map