Assay Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149441.2 | XP_015004927.1 | 1688 | 102 | ||
| XM_015149442.2 | XP_015004928.1 | 1511 | 102 | |||
| XM_015149444.2 | XP_015004930.1 | 1271 | 102 | |||
| XM_015149445.2 | XP_015004931.1 | 1218 | 102 | |||
| XM_015149446.2 | XP_015004932.1 | 1167 | 102 | |||
| XM_015149447.2 | XP_015004933.1 | 1201 | 102 | |||
| XM_015149448.2 | XP_015004934.1 | 1711 | 102 | |||
| XM_028828220.1 | XP_028684053.1 | 1090 | 102 | |||
| XM_028828221.1 | XP_028684054.1 | 1128 | 102 | |||
| XM_028828222.1 | XP_028684055.1 | 1124 | 102 | |||
| XM_028828223.1 | XP_028684056.1 | 1001 | 102 | |||
| GenBank mRNA | JU327141.1 | - | 1184 | 102 | ||
| JV071945.1 | - | 1347 | 102 | |||
| JV723868.1 | - | 1390 | 102 | |||
More Information
Set Membership: |
|

PANK2
Genomic Map