Assay Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265646.2 | NP_001252575.1 | 420 | 66 | ||
| XM_015136079.1 | XP_014991565.1 | 516 | 66 | |||
| XM_028846863.1 | XP_028702696.1 | 415 | 66 | |||
| XM_028846864.1 | XP_028702697.1 | 377 | 66 | |||
| GenBank mRNA | JU324022.1 | - | 421 | 66 | ||
| JV063095.1 | - | 495 | 66 | |||
| JV584183.1 | - | 510 | 66 | |||
| JV667413.1 | - | 420 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CUTA
Genomic Map