Assay Details
| Gene Symbol: ANGPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266116.1 | NP_001253045.1 | 1372 | 119 | ||
| XM_015145866.2 | XP_015001352.1 | 1528 | 119 | |||
| XM_028852125.1 | XP_028707958.1 | 760 | 119 | |||
| GenBank mRNA | JU320094.1 | - | 1372 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANGPT1
Genomic Map