Assay Details
| Gene Symbol: SMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126462.2 | XP_014981948.1 | 1944 | 59 | ||
| XM_015126463.2 | XP_014981949.1 | 2172 | 59 | |||
| XM_028841549.1 | XP_028697382.1 | 1929 | 59 | |||
More Information
Set Membership: |
|

SMPD3
Genomic Map