Assay Details
| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115506.2 | XP_014970992.2 | 3548 | 71 | ||
| XM_015115508.2 | XP_014970994.2 | 2587 | 71 | |||
| XM_015115509.2 | XP_014970995.2 | 1950 | 71 | |||
| GenBank mRNA | JU494868.1 | - | 212 | 71 | ||
| JV725127.1 | - | 166 | 71 | |||
More Information
Set Membership: |
|

MAML2
Genomic Map