Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148448.2 | XP_015003934.1 | 1188 | 76 | ||
| GenBank mRNA | JV070455.1 | - | 524 | 76 | ||
| JV636360.1 | - | 747 | 76 | |||
More Information
Set Membership: |
|

WNT2B
Genomic Map