Assay Details
| Gene Symbol: KYAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144487.2 | XP_014999973.2 | 747 | 89 | ||
| XM_015144489.2 | XP_014999975.2 | 812 | 89 | |||
| GenBank mRNA | JU335483.1 | - | 778 | 89 | ||
| JU470567.1 | - | 757 | 89 | |||
| JV047671.1 | - | 733 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KYAT3
Genomic Map