Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257661.1 | NP_001244590.1 | 870 | 65 | ||
| XM_015131175.1 | XP_014986661.1 | 969 | 65 | |||
| GenBank mRNA | JU322234.1 | - | 870 | 65 | ||
| JU476134.1 | - | 784 | 65 | |||
| JV717642.1 | - | 864 | 65 | |||
More Information
Set Membership: |
|

SNRK
Genomic Map