Assay Details
| Gene Symbol: KCNH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267017.1 | NP_001253946.1 | 376 | 67 | ||
| XM_015116791.2 | XP_014972277.1 | 396 | 67 | |||
| GenBank mRNA | JU471131.1 | - | 376 | 67 | ||
| JU471132.1 | - | 376 | 67 | |||
More Information
Set Membership: |
|

KCNH1
Genomic Map