Assay Details
| Gene Symbol: ACTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261802.2 | NP_001248731.1 | 659 | 72 | ||
| XM_015123793.2 | XP_014979279.1 | 784 | 72 | |||
| XM_015123794.2 | XP_014979280.1 | 784 | 72 | |||
| XM_015123795.2 | XP_014979281.1 | 784 | 72 | |||
| XM_015123796.2 | XP_014979282.1 | 784 | 72 | |||
| GenBank mRNA | JU336526.1 | - | 705 | 72 | ||
| JU336527.1 | - | 705 | 72 | |||
| JU477340.1 | - | 713 | 72 | |||
| JV048330.1 | - | 659 | 72 | |||
More Information
Set Membership: |
|

ACTN4
Genomic Map