Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032956.1 | NP_001028128.1 | 165 | 57 | ||
| GenBank mRNA | AY821556.1 | - | 165 | 57 | ||
| JU336137.1 | - | 400 | 57 | |||
| JV694854.1 | - | 426 | 57 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMP3
Genomic Map