Assay Details
| Gene Symbol: FADS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194717.3 | NP_001181646.1 | 1131 | 103 | ||
| XM_028832863.1 | XP_028688696.1 | 1602 | 103 | |||
| GenBank mRNA | JU337172.1 | - | 1103 | 103 | ||
| JU477899.1 | - | 1116 | 103 | |||
| JV048746.1 | - | 1040 | 103 | |||
| JV598799.1 | - | 1044 | 103 | |||
More Information
Set Membership: |
|

FADS2
Genomic Map