Assay Details
| Gene Symbol: ISY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094091.4 | XP_001094091.1 | 2069 | 65 | ||
| GenBank mRNA | JU321977.1 | - | 565 | 65 | ||
| JU478241.1 | - | 641 | 65 | |||
| JV588337.1 | - | 612 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ISY1
Genomic Map