Assay Details
| Gene Symbol: METAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261513.1 | NP_001248442.1 | 1073 | 73 | ||
| GenBank mRNA | JU322241.1 | - | 1076 | 73 | ||
| JU474850.1 | - | 1023 | 73 | |||
| JV046537.1 | - | 1073 | 73 | |||
| JV636651.1 | - | 1095 | 73 | |||
More Information
Set Membership: |
|

METAP1
Genomic Map