Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086798.4 | XP_001086798.4 | 1183 | 57 | ||
| XM_015110384.2 | XP_014965870.2 | 1231 | 57 | |||
| XM_015110385.2 | XP_014965871.2 | 1036 | 57 | |||
| GenBank mRNA | JU475983.1 | - | 1123 | 57 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAP3K20
Genomic Map