Assay Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094512.4 | XP_001094512.2 | 840 | 73 | ||
| XM_028837836.1 | XP_028693669.1 | 841 | 73 | |||
| GenBank mRNA | JU334094.1 | - | 610 | 73 | ||
| JU334095.1 | - | 610 | 73 | |||
More Information
Set Membership: |
|

CDH19
Genomic Map